Copper(I) sulfide, 99.5% (metals basis), Alfa Aesar

For Methods: Luminous Paints, Electrodes, Solar Cells

Manufacturer: Alfa Aesar 14718A1

Quantity 1kg

 View more versions of this product

Catalog No. AA14718A1

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 22205-45-4
    Physical Form Powder
    Assay Percent Range 99.5% (Metal basis)
    Molecular Formula Cu2S
    Formula Weight 159.14
    MDL Number MFCD00016062
    Chemical Name or Material Copper(I) sulfide

    Specifications

    Formula Weight 159.14
    Quantity 1kg
    Color Black
    EINECS Number 244-842-9
    Physical Form Powder
    MDL Number MFCD00016062
    Odor Odorless
    Assay Percent Range 99.5% (Metal basis)
    Molecular Formula Cu2S
    Appearance Black powder
    CAS 22205-45-4
    Chemical Name or Material Copper(I) sulfide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit