Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper(I) thiophene-2-carboxylate

Click to view available options
Quantity:
1g
5g
68986-76-5
C5H3CuO2S
190.683
MFCD02183524
SFJMFSWCBVEHBA-UHFFFAOYSA-M
copper i thiophene-2-carboxylate,cutc,unii-7519wbl07l,copper i thiophenecarboxylate,copper i thiophene-2-carboxylate,copper 2-thienylcarboxylate,cuprous 2-thienylcarboxylate,copper 2-thiophenecarboxylate,copper 1+ thiophene-2-carboxylate,copper i thiophene-2-carboxylate mi
11194830
copper(1+);thiophene-2-carboxylate
C1=CSC(=C1)C(=O)[O-].[Cu+]
68986-76-5
C5H3CuO2S
190.683
MFCD02183524
SFJMFSWCBVEHBA-UHFFFAOYSA-M
copper i thiophene-2-carboxylate,cutc,unii-7519wbl07l,copper i thiophenecarboxylate,copper i thiophene-2-carboxylate,copper 2-thienylcarboxylate,cuprous 2-thienylcarboxylate,copper 2-thiophenecarboxylate,copper 1+ thiophene-2-carboxylate,copper i thiophene-2-carboxylate mi
11194830
copper(1+);thiophene-2-carboxylate
C1=CSC(=C1)C(=O)[O-].[Cu+]
Applications
Reactant or reagent used for studies of xenobiotic response safener derivatives, synthesis of functionalized BODIPY dye analogs, orthogonal cross-coupling reactions, preparation of parent borondipyrromethene system and Copper mediated cross-coupling.

Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air. Keep container tightly closed. Store away from oxidizing agent.
Formula Weight 190.68
Physical Form Powder
Sensitivity Air & Light Sensitive
Color Brown to Red
Quantity 5g
Chemical Name or Material Copper(I) thiophene-2-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning