Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(I) thiophene-2-carboxylate, 90%, may contain approx. 20 wt.% Copper(I) oxide

Catalog No. AC430010010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430010010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430010010 Supplier Thermo Scientific Chemicals Supplier No. 430010010
Only null left
Add to Cart

CAS: 68986-76-5 | C5H3CuO2S | 190.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68986-76-5
Molecular Formula C5H3CuO2S
Molecular Weight (g/mol) 190.69
MDL Number MFCD02183524
InChI Key SFJMFSWCBVEHBA-UHFFFAOYSA-M
Synonym copper i thiophene-2-carboxylate, cutc, unii-7519wbl07l, copper i thiophenecarboxylate, copper 2-thienylcarboxylate, cuprous 2-thienylcarboxylate, copper 2-thiophenecarboxylate, copper 1+ thiophene-2-carboxylate, copper i thiophene-2-carboxylate mi
PubChem CID 11194830
IUPAC Name copper(1+);thiophene-2-carboxylate
SMILES C1=CSC(=C1)C(=O)[O-].[Cu+]

Specifications

Color Yellow
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 29.33 to 37.32% (Cu) ICP-OES
Merck Index 15,2506
Formula Weight 190.69
Percent Purity 90%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Copper(I) thiophene-2-carboxylate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.