Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) acetylacetonate, 98%

Catalog No. AC110651000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110651000 100 g Glass bottle
AC110650250 25 g Glass bottle
AC110655000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110651000 Supplier Thermo Scientific Chemicals Supplier No. 110651000
Add to Cart

CAS: 13395-16-9 | C10H14CuO4 | 261.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13395-16-9
Molecular Formula C10H14CuO4
Molecular Weight (g/mol) 261.76
MDL Number MFCD00000016
InChI Key QYJPSWYYEKYVEJ-FDGPNNRMSA-L
Synonym 2, 4-Pentanedione, metal derivative, Cupric acetylacetonate
IUPAC Name copper(2+) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES [Cu++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 245.0°C
Color Blue to Blue-Gray
Boiling Point 160.0°C (9.8 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Fieser 02,81; 03,62
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: 0.09g/100g alcohol (20°C),0.07g/100g benzene (20°C),readily soluble in chloroform,soluble in acetylacetone,dcm,dmf
Sublimation Point 160.0 °C
Formula Weight 261.76
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Copper(II) acetylacetonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 236-477-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.