Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(Ii) Chloride Dihydrate, ≥99.0%, ACS Reagent, Solstice

Catalog No. 60045852 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-852 500 g Poly Bottle
60-045-851 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-852 Supplier Solstice Supplier No. 307483500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Boiling Point 993°C
Melting Point 598°C
Physical Form Crystalline
Quantity 500 g
Packaging Poly Bottle
Density 2.53g/cm3 Vapor Density: >1 (vs air)
Linear Formula CuCl2 · 2H2O
UN Number UN2802
IUPAC Name copper(2+) dihydrate dichloride
Formula Weight 170.48g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Chemical Name or Material Copper(Ii) Chloride Dihydrate
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338
Precautionary Statement H290-H302 + H312-H315-H318-H410
DOTInformation 8/PGIII
EINECSNumber 231-210-2
RTECSNumber GL7030000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.