Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Copper(II) chloride hydrate, Puratronic™, 99.999% (metals basis)

Click to view available options
Quantity:
5g
25g
100g
10125-13-0
Cl2CuH4O2
170.476
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
coppertrace,copper ii chloride dihydrate,copper chloride dihydrate,caswell no. 235a,copper 2+ chloride dihydrate,cupric chloride usp,copper chloride cucl2 , dihydrate,epa pesticide chemical code 023701
61482
CHEBI:86318
dichlorocopper;dihydrate
O.O.Cl[Cu]Cl
10125-13-0
Cl2CuH4O2
170.476
MFCD00149674
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
coppertrace,copper ii chloride dihydrate,copper chloride dihydrate,caswell no. 235a,copper 2+ chloride dihydrate,cupric chloride usp,copper chloride cucl2 , dihydrate,epa pesticide chemical code 023701
61482
CHEBI:86318
dichlorocopper;dihydrate
O.O.Cl[Cu]Cl
UN Number UN2802
Merck Index 14,2633
Solubility Information Soluble in water.
Formula Weight 134.45 (Anhydrous)
Physical Form Crystalline Aggregates
Percent Purity 99.999%
Grade Puratronic™
Density 2.510 g/mL
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Color Blue-Green
Melting Point 100°C (decomposition)
pH 3.0 to 3.8
Quantity 5g
Chemical Name or Material Copper(II) chloride hydrate
Assay (metals basis)
Show More Show Less

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H318

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: COPPER CHLORIDE

EINECSNumber : 231-210-2

RTECSNumber : GL7030000

TSCA : Yes

Recommended Storage : Ambient temperatures