Learn More
Sigma Organic Chemistry copper ii i-butyrate | 500mg | MFCD00058695
Supplier: Sigma Organic Chemistry CDS003339500MG

copper ii i-butyrate , 500mg
About this Item:
MDL #: MFCD00058695
Molecular Weight: 237.74
UNSPSC Code: 12352200
Chemical Formula: C8H14CuO4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.