Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) nitrate hemi(pentahydrate), 98%

Catalog No. AAA140040I
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA140040I 5000 g
AAA1400430 250 g
AAA140040B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA140040I Supplier Thermo Scientific Chemicals Supplier No. A140040I
Add to Cart

CAS: 19004-19-4 | CuN2O6·2.5H2O

Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) nitrate hemi(pentahydrate) is used for the catalytic oxidative coupling of 2,6-dimethylphenol to produce a polymer which is an important engineering material. Its solutions are used in textiles and polishing agents for other metals. It is commonly used in school laboratories to demonstrate chemical voltaic cell reactions.

Solubility
Soluble in water, ammonia and ethanol

Notes
Incompatible with reducing agents, organic materials and powdered metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19004-19-4
Molecular Formula CuN2O6·2.5H2O
MDL Number MFCD00149671
Synonym dicopper tetranitrate pentahydrate, copper ii nitrate hemipentahydrate, cupric nitrate, hydrate, copper, reference standard solution, copper nitrate hemipentahydrate, cupric nitrate hemipentahydrate, 2cu.4no3.5h2o, cupric nitrate hemi pentahydrate

Specifications

Boiling Point 170°C (decomposition)
Melting Point ∼115°C
Quantity 5000 g
Linear Formula Cu(NO3)2·2.5H2O
UN Number UN3085
Merck Index 14,2643
Solubility Information Soluble in water,ammonia and ethanol
Formula Weight 232.59 (187.55 Anhydrous)
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.32
Chemical Name or Material Copper(II) nitrate hemi(pentahydrate)
Show More Show Less
Hazard Category H272-H314-H335
Hazard Statement GHS H Statement
H272-H314-H318-H302
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 221-838-5
RTECSNumber GL7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.