Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. AA1069914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1069914 25 g
AA1069906 5 g
AA1069922 100 g
AA1069936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1069914 Supplier Thermo Scientific Chemicals Supplier No. 01069914
Add to Cart

CAS: 13778-31-9 | CuN2O6 | 187.55 g/mol

It serves as a catalyst for oxidative coupling of 2,6-dimethylphenol, resulting in an important engineering polymer material. Also used in textiles and polishing agents for other metals. It is a precursor for copper oxide, which finds use as a catalyst for many organic transformations. Menke conditions employ copper nitrate for aromatic nitrations. Claycop reagent, the hydrated copper nitrate deposited on clay, finds use in many organic reactions, for example, oxidation of dithioacetals to carbonyls and thiols to disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13778-31-9
Molecular Formula CuN2O6
Molecular Weight (g/mol) 187.55
MDL Number MFCD00149669
InChI Key XTVVROIMIGLXTD-UHFFFAOYSA-N
Synonym copper ii nitrate hydrate, copper nitrate hydrate, acmc-20aldm, cupric nitrate hydrate, cu.2no3.h2o, copper 2+ ion hydrate dinitrate, copper 2+ hydrate dinitrate, copper ii nitrate hydrate, puratronic, copper 2+ nitrate-water 1/2/1, nitric acid, copper 2+ salt, hydrate 8ci,9ci
PubChem CID 15303652
SMILES [Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Blue
Physical Form Crystalline Aggregates
Quantity 25 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in ethanol and water. Insoluble in ethyl acetate.
IUPAC Name copper;dinitrate;hydrate
Formula Weight 187.55 (anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Copper(II) nitrate hydrate
Hazard Category H272-H314-H335
Precautionary Statement P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 221-838-5
RTECSNumber GL7875000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.