Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) pyrithione

Catalog No. AC457210010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457210010 1 g
AC457210050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457210010 Supplier Thermo Scientific Chemicals Supplier No. 457210010
Add to Cart

CAS: 14915-37-8 | C10H8CuN2O2S2 | 315.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14915-37-8
Molecular Formula C10H8CuN2O2S2
Molecular Weight (g/mol) 315.86

Specifications

Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 315.86
Percent Purity ≥97.5% (HPLC)
Infrared Spectrum Authentic
Chemical Name or Material Copper(II) pyrithione
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated cl
EINECSNumber 238-984-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.