Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) Sulfate, Pentahydrate, ACS, 99.0 to 100.5%, MilliporeSigma™

Catalog No. M1027901000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1027901000 1 kg
M1027900250 250 g
M1027905000 5 kg
M1027909050 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1027901000 Supplier MilliporeSigma Supplier No. 1.02790.1000
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 99.0 to 100.5%
Linear Formula CuSO4 · 5H2O
Formula Weight 249.68
Grade ACS/EP/ISO/Reagent
Density 2.284g/cm3 (20°C)
Chemical Name or Material Copper(II) Sulfate, Pentahydrate, EMSURE™, For Analysis
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.