Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Copper (II) Sulfate Pentahydrate, Reagent Grade
SCI_ED

Catalog No. S25286B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Reagent Grade - Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25286B 3 kg Reagent Grade - Powder
S25286 100 g Reagent Grade - Powder
S25286A 500 g Reagent Grade - Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25286B
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
Quantity 3 kg
Linear Formula CuSO4·5H2O
Formula Weight 249.68
Packaging Reagent Grade - Powder
Density 2.2840g/cm3
Chemical Name or Material Copper (II) Sulfate, Pentahydrate, Reagent Grade
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 6.1, III, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.