Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Manganese Nickel wire, 0.1mm (0.004 in.) dia., Thermo Scientific™

Catalog No. AA43188H4
Change view
Click to view available options
Quantity:
100 m
500 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43188H4 100 m
AA43188H9 500 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43188H4 Supplier Thermo Scientific Chemicals Supplier No. 43188H4

Resistance alloy for thermocouples and electronics industry

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Manganese Nickel wire
Name Note 0.1mm (0.004 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Cu:Mn:Ni; 84:12:4 wt%
Quantity 100 m
Weight ≈0.066g/m
Form Wire
MDL Number MFCD01866121
Synonym Manganin™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.