Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Metal, Shot, ACS, 99.9%, Spectrum™ Chemical
SDP

Catalog No. 18609937 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-609-937 125 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-609-937 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C1696125GM

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
IUPAC Name copper
SMILES [Cu]

Specifications

CAS Max % 1
Quantity 125 g
Packaging Poly Bottle
Form Shot
Percent Purity 99.9%
Grade ACS
Chemical Name or Material Copper Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.