Learn More
Sigma Organic Chemistry Copper phthalocyanine-3,4',4",4"'-tetrasulfonic acid tetrasodium salt | 1G | 123439-80-5
Supplier: Sigma Organic Chemistry 2453561G
            Copper phthalocyanine-3,4',4",4"'-tetrasulfonic acid tetrasodium salt, 1G
About This Item:
Linear Formula: C32H12CuN8O12S4 -+ 4Na
UNSPSC Code: 12352103
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.