Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Plasma Grade Reference Standard, 1mL = 1mg Cu, 1000ppm Cu, Ricca Chemical
SDP

Catalog No. 23014 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23014 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23014 Supplier Ricca Chemical Company Supplier No. 23014

Chemical Identifiers

CAS 7440-50-8 , 7697-37-2 , 7732-18-5
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Packaging Poly natural
Concentration or Composition Notes 1000ppm Cu (Cu in 5% HNO3)
Equivalence 1 mL = 1 mg Cu
Physical Form Liquid
Color Blue
Quantity 120 mL
Chemical Name or Material Copper Plasma Grade Reference Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.