Alfa Aesar™ Copper powder, -100 mesh, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 3968206

Quantity 5g

 View more versions of this product

Catalog No. AA3968206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-50-8
    MDL Number MFCD00010965
    UN Number UN3089
    Chemical Name or Material Copper powder

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 7440-50-8
    Mesh Size −100 Mesh
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00010965
    UN Number UN3089
    Odor Odorless
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228-H319-H335
    Flammable solid.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color Red brown
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    RTECS Number GL5325000
    Quantity 5g
    Chemical Name or Material Copper powder
    Assay 99.999%, (metals basis)

    GHS H Statement
    H228-H319-H335
    Flammable solid.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit