Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper (Shot/Certified ACS), Fisher Chemical

Catalog No. C430500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C430-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C430-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Color Red Brown
Melting Point 1083°C
Boiling Point 2595°C
Atomic Weight 63.54
Density 8.92g/cm³
Quantity 500 g
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 1mmHg at 1628°C
Form Solid
Assay Percent Range ≥99.9 %
Formula Weight 63.55
Percent Purity ≥99.9%
Grade Certified ACS
Chemical Name or Material Copper
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.