Learn More
Copper Sulfate Solution, 1.6% with 0.6% Sulfuric Acid, Reagents
Specifications
Specifications
| Chemical Name or Material | Copper Sulfate Solution, 1.6% with 0.6% Sulfuric Acid |
| Boiling Point | 100°C |
| Color | Blue |
| CAS | 7732-18-5, 7758-99-8 |
| pH | <2 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Molecular Formula | CuSO4/H2O/H2SO4 |
| Synonym | Copper(II) Sulfate |
| Percent Purity | 1.6% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.