Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate Solution, Specific Gravity 1.186, for UL Corrosion Test, Ricca Chemical
SDP

Catalog No. 23481 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
23481 4 L Natural Poly Bottle
234832 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23481 Supplier Ricca Chemical Company Supplier No. 23481
Add to Cart
Add to Cart

For UL Corrosion Test

Chemical Identifiers

CAS 7758-99-8 , 7732-18-5 , 1317-38-0
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Min % 22.62, 75.0, 0.03
CAS Max % 23.54, 78.0, 0.03
Grade Laboratory
Packaging Natural Poly Bottle
pH 3 to 3.6
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Copper Sulfate Solution, Specific Gravity 1.186
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.