Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate-Sulfamic Acid Inhibitor, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88554 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88554 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88554 Supplier Aqua Phoenix Scientific Supplier No. CU7975Q
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical reagent for laboratory applications, featuring induction seals to ensure product integrity.

Specifications

Chemical Name or Material Copper Sulfate
CAS 64-19-7, 7732-18-5
Physical Form Liquid
Quantity 1 L
Packaging Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.