Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Coprostanol, Cholesterol Derivative, >98% Purity, 25 mg

Catalog No. 502706068
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6068 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6068 Supplier Abcam Supplier No. AB14388225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biomarker for vertebrate intestinal microbe populations derived from the bacterial biohydrogenation of cholesterol.

  • Molecular Weight: 388.7 Da
  • Purity: >98%
  • Description: Cholesterol (ab142501) derivative; forms from bacterial biohydrogenation of cholesterol
  • Application: Biomarker for vertebrate intestinal microbe populations in vivo
  • Composition: 100% Coprostanol
  • CAS Number: 360-68-9

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Cholesterol
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Buffer 100% Coprostanol
Content And Storage Store at -20°C.
Quantity 25 mg
For Use With (Application) Biomarker for vertebrate intestinal microbe populations
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.