Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COPS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7088001 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6326 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6326 Supplier Proteintech Group Inc Supplier No. 104641AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSN4 is a subunit of the JUN activation-domain binding protein 1 (JAB1), containing signalosome complex (or COP9 complex) which has kinase activity and phosphorylates C-JUN. The signalosome complex may play a role in signal transduction.

Specifications

Antigen COPS4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene COPS4
Gene Accession No. O88544, Q68FS2, Q9BT78
Gene Alias COPS4, CSN4, SGN4, Signalosome subunit 4
Gene Symbols COPS4
Host Species Rabbit
Immunogen COPS4 Fusion Protein Ag0735
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26891, 360915, 51138
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.