Learn More
TARGETMOL CHEMICALS INC COPTISINE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5S0053
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Coptisine (Coptisin) treatment increases cell viability based on its reversal effect on the enhanced activity of Indoleamine 2 3-dioxygenase . 2. Coptisine treats myocardial I/R likely through suppressing myocardial apoptosis and inflammation by inhibiting the Rho/ROCK pathway. 3. Coptisine is a potential anti-osteosarcoma drug candidate via exerting a strong anti-osteosarcoma effect with very low toxicity . 4. Coptisine with a high dosage could inhibit cholesterol synthesis via suppressing the HMGCR expression and promoting the use and excretion of cholesterol via up-regulating LDLR and CYP7A1 expression. 5. Coptisine suppresses adhesion migration and invasion of MDA-MB-231 breast cancer cells in vitro the down-regulation of MMP-9 in combination with the increase of TIMP-1 possibly contributing to the anti-metastatic function for breast cancer. purity: 99%
Catalog No. 50-257-6652
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.