Learn More
Proteintech Group Inc CoraLite488-conjugated MLKL Monoclonal antibody
SDP
Supplier:  Proteintech Group Inc CL48866675100UL
Encompass_Preferred
The MLKL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MLKL. This antibody recognizes human, mouse antigen. The MLKL antibody has been validated for the following applications: IF analysis. This antibody is coralite488.
Catalog No. 50-225-7233
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.