Learn More
VAI™ Core2Clean System™ Spare Pressure Output Hose
Catalog No. 19546801 Shop All VAI Products
Change view
Click to view available options
Overall Length (Metric):
2.43 m
4.5 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Overall Length (Metric)
19-546-801 2.43 m
01-549-878 4.5 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-546-801 Supplier VAI™ Supplier No. C2C1004BS
  • $3,901.00
    $3,543.65 / Each of 1
    Save $357.35
    9% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
The Core2Clean™ Plus Systems were created by Veltek Associates, Inc., to provide specialized cleaning equipment for use in controlled environments.
  • Regulator, complete unit and fittings for regulating Core2Clean incoming air pressure
  • Pressure Relief Valve Assembly, automatic safety relief valve, factory set to prevent over pressurization of Core2Clean tank
  • Stainless Steel
Specifications
Product Type Pressure Output Hose
For Use With (Equipment) Core2Clean Plus Systems
Overall Length (Metric) 2.43 m
Overall Length (English) 8 ft.