Learn More
VAI Core2Write™ Labels
Catalog No. 19546595 Shop All VAI Products
Change view
Click to view available options
Size (English):
2 x 1 in.
4 x 3 in.
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Sterility
19-546-595 2 x 1 in. Sterile
19-546-594 4 x 3 in. Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-546-595 Supplier VAI Supplier No. C2WR4X3BLK02
Add to Cart
Add to Cart
Request a Quote
Core2Write™ Labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures.
  • Abrasion, tear resistant, and smear resistant
  • Lightweight and flexible but exceptionally durable
  • Inks adhere and dry immediately
Specifications
Format Roll
Sterility Sterile
Quantity 1000 Labels/Roll, 4 Rolls/Cs.
Size (English) 2 x 1 in.
Size (Metric) 51 x 25 mm