Learn More
Horizon Scientific Corepoint™ Standard Flammable Storage Freezer with Natural Refrigerants
SDP
Catalog No. 16998544 Shop All Nor-Lake Scientific Products
Change view
Click to view available options
Temperature Range:
-15°C to -25°C
Catalog No. Temperature Range
16-998-544 -15°C to -25°C
1 options
Catalog No. 16-998-544 Supplier Horizon Scientific Supplier No. CPSHCFFS30
Add to Cart
Add to Cart
This freezer offers a secure solution for storing volatile materials in laboratory and industrial settings.
  • Meets NFPA and OSHA guidelines for hazardous work environments 45 and 70 for the refrigerated storage of flammable materials
  • Powered by Microprocessor temperature technology for precise and stable temperature control
  • Offers EPA SNAP compliant hydrocarbon refrigerants
  • Equipped with audible and visual alarms to safeguard stored contents
  • Offers low energy consumption and high safety standards
Specifications
Type Flammable Freezer
Capacity (English) 30 cu.ft.
Capacity (Metric) 850 L
Temperature Range -15°C to -25°C
Door Style Solid Swing
No. of Doors 1
Cabinet Material High-impact, Powder-coated Paint
Chamber Material Powder-coated Steel
Shelves 4
Certifications/Compliance cULus, NFPA 45 and 70, EPA SNAP
Style Upright
Monitoring Options Audible and Visual High and Low Temperature Alarms
Plug Type NEMA 5-15 P
Voltage 110/120 V
Compressor Bottom-mounted