Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Horizon Scientific Corepoint™ Standard Hazardous Location Freezer with Natural Refrigerants
SDP

Catalog No. 16998543 Shop All Nor-Lake Scientific Products
Change view
Click to view available options
Temperature Range:
-15°C to -25°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range
16-998-543 -15°C to -25°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-998-543 Supplier Horizon Scientific Supplier No. CPSHCEFS30
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This freezer meets NFPA and OSHA guidelines for the safe storage of materials in potentially hazardous work environments.

  • EPA SNAP compliant hydrocarbon refrigerants
  • Microprocessor temperature technology for precise and stable temperature control
  • Low energy consumption and high safety standards
  • Please note: This unit must be hardwired for safety

Specifications

Type Hazardous Freezer
Capacity (English) 30 cu.ft.
Capacity (Metric) 850 L
Temperature Range -15°C to -25°C
Door Style Solid Swing
No. of Doors 1
Cabinet Material Coated Steel
Chamber Material Painted Steel
Shelves 4
Certifications/Compliance UL listed to U.S. and Canadian requirements, NFPA, OSHA, EPA SNAP
Style Upright
Monitoring Options Audible and Visual High and Low Temperature Alarms
Plug Type No Plug, Hardwired
Voltage 110/120 V
Compressor Bottom-mounted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.