Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ CoreShield™ 15-Gauge Nylon ANSI A1 Cut-Resistant Gloves, MicroFoam Nitrile Coated

Catalog No. 19085657
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-085-657 X-Large
19-085-726 Large
19-085-655 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Catalog No. 19-085-657 missing translation for 'mfr' Honeywell™ missing translation for 'supplierNo' 211515B/10XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight, cut-resistant nylon lined gloves provide comfort and prevent hand fatigue for workers who need to wear them all day long.

  • Lightweight, cut-protective gloves help to prevent hand fatigue for workers who need to wear them all day long
  • High-performance coating is liquid repellent to increase dry grip performance
  • CoreShield™ engineered yarn is skin friendly and comfortable to shield hands from the cut resistant material
  • Special abrasion technology provides twice the durability
  • Hand and wrist protection with thumb reinforcement
  • Touchscreen resistive and capacitive
  • Launderable, with no significant changes in the glove performance or size after 5 washing cycles
  • Certified STANDARD 100 by OEKO-TEX™

Specifications

ANSI Abrasion Rating ANSI 6
Coating Material Nitrile Foam
Color Black, Red
Cut Resistance Rating ANSI A1
Hand Specific
Material Nylon
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Manufacturing, automotive, metal stamping, glass handling, construction and logistics
Glove Cuff Style Knit
Model 21-1515B/10XL
Packaging Type Plastic Bag
Series CoreShield™
Yarn Gauge 15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.