Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ CoreShield™ HPPE ANSI A2 Cut-Resistant Gloves, MicroFoam Nitrile Coated

Catalog No. 19085666 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-085-666 X-Large
19-085-664 Medium
19-085-665 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-085-666 Supplier Honeywell™ Supplier No. 227513B/10XL
  • $21.25
    $9.65 / Pair

    Save $11.60

    55% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $2,106.00
    $967.65 / Case of 100 PR

    Save $1,138.35

    54% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight, cut-resistant gloves feature CoreShield™ engineered yarn to provide comfort and to prevent hand fatigue for workers who need to wear them all day long.

  • Lightweight, cut-protective gloves help to prevent hand fatigue for workers who need to wear them all day long
  • High-performance coating is liquid repellent to increase dry grip performance
  • CoreShield™ engineered yarn is skin friendly and comfortable to shield hands from the cut resistant material
  • Special abrasion technology provides twice the durability
  • Hand and wrist protection with thumb reinforcement
  • Touchscreen resistive and capacitive
  • Launderable, with no significant changes in the glove performance or size after 5 washing cycles
  • Certified STANDARD 100 by OEKO-TEX™

Specifications

ANSI Abrasion Rating ANSI 6
Coating Material Nitrile Foam
Color Black, Yellow
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size X-Large
For Use With (Application) Manufacturing, automotive, metal stamping, glass handling, construction and logistics
Glove Cuff Style Knit
Model 22-7513B/10XL
Packaging Type Plastic Bag
Series CoreShield™
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.