Learn More
Honeywell - PPE CoreShield™ Double 13G WL Glove, 3/4 Nitrile (Micro-Foam) Coating
Catalog No. 19066902 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-066-902 Large
19-066-901 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-066-902 Supplier Honeywell - PPE Supplier No. 230D23W9/L
Add to Cart
Add to Cart
With the Honeywell CoreShield Double gloves, cut protection in oily environments is transformed — no water or oil on hands, reduced sweat and bad odors, less bacteria on the gloves; just enhanced grip and comfort.
  • Provides superior cut protection
  • Offer efficient grip in extremely greasy environments and improved comfort and dexterity for a wide range of applications
  • For use in machining operations, automobile assemblies, food handling applications, and manufacturing industries
Specifications
Construction Single Knit
Coating Material Nitrile MicroFoam
Color Black
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free Yes
Size Large
Gender Unisex
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 3
Thumb Style Wing
Yarn Gauge 13 Gauge