Learn More
Honeywell - PPE CoreShield™ Double 18G BB Glove, 3/4 Nitrile (Micro-Foam) Coating
Catalog No. 19066907 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Catalog No. Size
19-066-907 Medium
1 options
Catalog No. 19-066-907 Supplier Honeywell - PPE Supplier No. 247D28B8/M
Add to Cart
Add to Cart
With the Honeywell CoreShield Double gloves, cut protection in oily environments is transformed — no water or oil on hands, reduced sweat and bad odors, less bacteria on the gloves; just enhanced grip and comfort.
  • Provides superior cut protection
  • Offer efficient grip in extremely greasy environments and improved comfort and dexterity for a wide range of applications
  • For use in machining operations, automobile assemblies, food handling applications, and manufacturing industries
Specifications
Construction Seamless Knit
Coating Material Nitrile MicroFoam
Color Black
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Size Medium
Gender Unisex
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Thumb Style Wing
Yarn Gauge 18 Gauge