Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Corey Lactone 98.0+%, TCI America™
SDP

Catalog No. H14181G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14181G 1 g
H1418200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14181G Supplier TCI America Supplier No. H14181G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32233-40-2
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00075234
InChI Key VYTZWRCSPHQSFX-GBNDHIKLSA-N
Synonym --corey lactone diol, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl hexahydro-2h-cyclopenta b furan-2-one, --corey lactone, corey lactone diol, --corey diol corey lactone, 3ar,4s,5r,6as---hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 1s,6s,5r,7r-7-hydroxy-6-hydroxymethyl-2-oxabicyclo 3.3.0 octan-3-one, 3ar,4s,5r,6as-hexahydro-5-hydroxy-4-hydroxymethyl-2h-cyclopenta b furan-2-one, 3ar,4s,5r,6as-5-hydroxy-4-hydroxymethyl-3,3a,4,5,6,6a-hexahydrocyclopenta b furan-2-one, l-corey lactone
PubChem CID 2724453
IUPAC Name (3aR,4S,5R,6aS)-5-hydroxy-4-(hydroxymethyl)-hexahydro-2H-cyclopenta[b]furan-2-one
SMILES OC[C@H]1[C@H](O)C[C@@H]2OC(=O)C[C@H]12

Specifications

Melting Point 116°C
Color White
Quantity 1 g
Formula Weight 172.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Corey Lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.