Learn More
Julabo USA Inc CORIO CD-1200F R1270 EA 230V
SDP
Supplier:  Julabo USA Inc 9012717.N1.13
Encompass_Preferred
Refrigerated/heating circulator, JULABO, CORIO CD-1200F, 230V/60Hz, R1270 Refrigerant. -40…+150C. 2kW heater. Cooling power 1.25kW at 20C 1.1kW at 0C 630W at -20C 380W at -30C 210W at -40C. Pump 16LPM 4.8 PSI. USB plus data-logging. M16x1 pump fittings with 8 and 12mm barbs. 5...7.5L bath tank. 13 x 18.5 x 27.6 in. 93lbs
Catalog No. NC2751009
Only null left
Add to Cart
Request a Quote
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov