Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Coriolis Effect Apparatus
SDPSCI_ED

Catalog No. S111029 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S111029 6 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S111029 Supplier United Scientific™ Supplier No. UNCOEF001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With this kit, students investigate how a spinning platform causes a moving object travel in a curved path.

This apparatus helps students to understand the Coriolis effect, the phenomenon whereby an object moving on a spinning surface will travel in a curved path. The demonstrations that follow will allow students to observe the Coriolis effect on a small scale, then explore how the Earth's constant rotation influences wind patterns and ocean currents around the world.

Components:

  • Turntable
  • Erasable Tracing Surface
  • Ramp
  • Steel Balls (2)

Specifications

Grade 6 to 14
Format Kit
Product Type Coriolis Effect
Languages English
Subject Earth Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.