Learn More
Pendotech Pendotech Coriolis Mass Flowmeter
Supplier:  Pendotech PCFM32
Encompass_Preferred
PendoTECH Coriolis Mass Flowmeter with 0.25” /6.35 mm ID (range 12 - 4,000 grams/min)
Catalog No. NC0851540
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.