Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 0.05% Trypsin/0.53mM EDTA in HBSS w/o Calcium and Magnesium
GSA_VA

Catalog No. MT25051CI Shop All Corning Products
Change view
Click to view available options
Quantity:
6 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT25051CI 6 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT25051CI Supplier Corning™ Supplier No. 25051CI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to enzymatically release adherent cells from tissue culture plates for passaging.

  • Contains trypsin at a concentration of 0.05% with 0.53mM EDTA, and sodium bicarbonate
  • Low endotoxin content
  • EDTA enhances the efficacy of trypsin

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
Without Additives No Calcium, No Magnesium
pH 7 to 7.6
Quantity 6 x 100 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.