Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Automation Pipette Tips for Molecular Devices FLIPR Tetra Workstation
GSA_VAGREENER_CHOICE

Catalog No. 14223556 Shop All Axygen Products
Change view
Click to view available options
Color:
Black
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-223-556 Clear
14-223-557 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-223-556 Supplier Axygen™ Supplier No. FT384CR
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Pipette tips manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality

  • 384 Tip format
  • Polypropylene
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Compatibility: Molecular Devices FLIPR Tetraworkstation
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 30 μL
    Sterility Non-sterile
    Color Clear
    Format Rack
    Tip Style Automation, Round
    For Use With (Equipment) Molecular Devices FLIPR Tetra Automation System, Hamilton MICROLAB STAR
    Material Polypropylene
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.