Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ BioCoat™ Fibronectin Rectangular Culture Flask
GREENER_CHOICE

Catalog No. 08774412 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
250 mL
750 mL
Culture Area:
175 cm2
75 cm2
Neck Style:
Canted
Straight
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Culture Area Neck Style
08-774-412 750 mL 175 cm2 Straight
08-774-65 250 mL 75 cm2 Canted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08-774-412 Supplier Corning™ Supplier No. 354526
Add to Cart
Add to Cart

Corning™ BioCoat™ products are the ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.

Human Fibronectin (HFN) is a widely distributed glycoprotein that is used as a substrate to promote attachment of cells through its central-binding domain RGD sequence. HFN is a product of most mesenchymal and epithelial cells and is present in both the ECM and plasma. The principal function of HFN appears to be in cellular migration during wound healing and development, regulation of cell growth and differentiation, and haemostasis/thrombosis.

  • Matrix components are applied as an even, optically clear coating
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells

Applications include:

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced-serum culture
  • Cell adhesion assays
  • Studies of effects of HFN on cell behavior
  • Improving survival of primary cells in culture

Source

  • Human plasma
  • Note: Source material tested for hepatitis B antigen and HIV-1 antibody

Quality Control

  • Tested for ability to promote attachment and spreading of BHK-21 hamster kidney cells
  • Tested and found negative for bacteria and fungi
  • Fibronectin purity >90% by SDS-PAGE
TRUSTED_SUSTAINABILITY

Specifications

Closure Material Phenolic
Capacity (Metric) 750 mL
Material Polystyrene
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Neck Style Straight
Shape Rectangular
Surface Coating Fibronectin
Sterility Sterile
Culture Area 175 cm2
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.