Learn More
Corning™ CellCube™ Tubing Adaptor
GREENER_CHOICE
Catalog No. 10320191 Shop All Corning Products
Change view
Click to view available options
Product Type:
4 AseptiQuik™ S Connectors, 4 Tubes Clamps
AseptiQuik™ S Connected to a Female MPC
AseptiQuik™ S Connected to a Male MPC
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
10-320-191 AseptiQuik™ S Connected to a Female MPC
10-320-190 4 AseptiQuik™ S Connectors, 4 Tubes Clamps
10-320-192 AseptiQuik™ S Connected to a Male MPC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-320-191 Supplier Corning™ Supplier No. 3237
Add to Cart
Add to Cart
A new closed system offering designed to help reduce the risk of adventitious contamination
  • Closed System Cell Cube
  • CellCube 10, 25, 100 modules, circulation loops, and connectors
  • Allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik™ and MPC connectors.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner Tubing 9.53 mm
Includes Thermoelastic Tubing, Aseptic Connector to Female MPC
No. per Case 4/Cs.
No. per Pack 1/Pk.
Product Type AseptiQuik™ S Connected to a Female MPC