Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X

Catalog No. MT21031LB Shop All Corning Products
Change view
Click to view available options
Quantity:
1 x 10,000 mL
1 x 20,000 mL
6 x 1000 mL
6 x 500 mL
Physical Form:
Liquid
Includes:
Calcium and Magnesium
No Calcium, No Magnesium
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Physical Form Includes
MT21031LB 1 x 20,000 mL Liquid No Calcium, No Magnesium
MT21030CM 6 x 1000 mL Liquid Calcium and Magnesium
MT21031LX 1 x 10,000 mL Liquid No Calcium, No Magnesium
MT21030CV 6 x 500 mL Liquid Calcium and Magnesium
MT21031CM 6 x 1000 mL Liquid No Calcium, No Magnesium
MT21031CV 6 x 500 mL Liquid No Calcium, No Magnesium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. MT21031LB Supplier Corning™ Supplier No. 21031LB
Add to Cart
Add to Cart
This item is not returnable. View return policy

With low endotoxin content.

Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 1 X
pH 7.3 to 7.5
Physical Form Liquid
Quantity 1 x 20,000 mL
Formulation Potassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Sodium Chloride 8g/L, Disodium Phosphate 1.15g/L
Includes No Calcium, No Magnesium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.