Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Gentamicin Sulfate

Catalog No. MT61098RA Shop All Corning Products
Change view
Click to view available options
Quantity:
1 x 100 mg
1 x 1 g
10 x 10 mL
Form:
Liquid
Powder
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MT61098RA 1 x 100 mg Powder
MT30005CR 10 x 10 mL Liquid
MT61098RF 1 x 1 g Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. MT61098RA Supplier Corning™ Supplier No. 61098RA
Add to Cart
Add to Cart
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Packaged in plastic bottles

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Form Powder
Product Type Antibiotic
Sterility Sterile
Format Plastic Bottle
Quantity 1 x 100 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.