Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Tryptic Soy Broth, Animal-Free, Powder

Catalog No. MT61411RO Shop All Corning Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MT61411RO 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MT61411RO Supplier Corning™ Supplier No. 61411RO
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For an accurate assessment of sterility throughout the manufacturing process

  • Produced under the highest industry standards to ensure superior results
  • Nonanimal origin
  • For microbiology
  • Clear, yellowish-brown appearance
  • Sterile, ready-to-use solution
  • Complies with USP requirements

For testing the sterility of animal-free biopharmaceutical processes.

Order Info

Shipped under ambient conditions.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance USP
Content And Storage 15°C to 30°C
Form Powder
Product Type Microbiology Media
Sterility Sterile
Formulation Nonanimal origin Peptone 20, Dextrose 2.5, NaCl 5, K2HPO4 2.5
For Use With (Application) For testing the sterility of animal-free biopharmaceutical processes
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.