Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CellSTACK™
GREENER_CHOICE

Catalog No. 10320195 Shop All Corning Products
Change view
Click to view available options
Culture Area:
1272 cm²
6360 cm²
Height (Metric):
31 mm
203 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Height (Metric)
10-320-195 1272 cm² 31 mm
10-320-196 6360 cm² 203 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 10-320-195 Supplier Corning™ Supplier No. 11874
Add to Cart
Add to Cart
Request a Quote
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Products meet a minimum Sterility Assurance Level (SAL) of 10-6

  • Product has been sterilized and dosimetrically released 
  • Products meet a minimum Sterility Assurance Level (SAL) of 10-6
  • Non-Pyrogenic
  • Bacterial Endotoxins test methodologies, routine monitoring, and alternative to batch testing
  • Tested for the attribute of cell attachment and growth utilizing an attachment-dependent mammalian cell line
TRUSTED_SUSTAINABILITY

Specifications

Description Nonpyrogenic
Height (Metric) 31 mm
Includes Custom Fill and Vent Accessories
Length (Metric) 335 mm
Width (Metric) 206 mm
Culture Area 1272 cm²
Tray Layers 2 Chamber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.