Learn More
Corning™ CrystalEX384-Well Flat Bottom Protein Crystallization Microplate
Catalog No. 07200820 Shop All Corning Products
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
Catalog No. Well Shape
07-200-820 Flat Bottom (F-well)
1 options
Catalog No. 07-200-820 Supplier Corning™ Supplier No. 3775
The only microplate designed for full automation in crystal screening
96-well Microplates
  • Features 96 large reservoir (reagent) wells and 96 corresponding protein wells
  • Conical bottom protein wells allow improved centering of the protein drop.
  • Compatible with manual pipettors and automation
  • Novel merged well design provides efficient vapor space for protein crystallization.
  • Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
384-well Microplate
  • Meets ANSI/SBS standard footprint dimensions for microplates
  • Ideal for fully automated crystal screening
  • Features 192 reservoir wells and 192 corresponding protein wells
  • Flat bottom protein wells are optimized for imaging of crystals.
  • Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Material Advanced Polymer
No. of Wells 384
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
Includes 192 reservoir wells and 192 corresponding protein wells
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Depth (Metric) 2.3 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov