Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ DeckWorks™ Low Binding Hinged Rack Pipette Tips

Catalog No. 10320728 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
10-320-728 1 to 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-320-728 Supplier Corning™ Supplier No. 4139
Add to Cart
Add to Cart

Used for quantitative assays in which sample binding can significantly alter results.

  • Low binding barrier tips feature an inert, high-density polyethylene barrier to eliminate aerosol carryover contamination during critical pipetting procedures.
  • Low binding barrier tips will not trap liquids or inhibit PCR in the event of over pipetting.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Filtered Filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Certifications/Compliance EN ISO 13485 compliant, BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, Human DNA Free, DNase/RNase Free, PCR Inhibition tested, Sterility compliant with ANSI/AAMI/ISO 11137, Quality Control Tested
Clean Claims Nonpyrogenic, Human DNA-free, RNase/DNase-free, Sterility Assurance Level 10−3
For Use With (Application) Used for quantitative assays in which sample binding can significantly alter results
Format Hinged Rack
Tip Style Low Retention, Round
Packaging Rack
Quantity 768 Tips/Pk.
Product Type Low Binding Barrier Pipette Tips
Includes Tips, Rack, Pack, Case
Graduated Yes
Product Line DeckWorks
Material HDPE
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.