Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Human Umbilical Vein Endothelial Cells (HUVEC-2)

Catalog No. 08774388 Shop All Corning Products
Change view
Click to view available options
Quantity:
>5 x 105 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-774-388 >5 x 105 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-774-388 Supplier Corning™ Supplier No. 354151

Used as model for studying endothelial differentiation and modulation of endothelial tube formation by antiangiogenic agents

Corning™ HUVEC-2 cells are qualified for use in Corning™ BioCoat™ Angiogenesis Systems: Endothelial Cell Migration. The cells may be used in Corning BioCoat Endothelial Cell Invasion and Tube Formation assays. Packaged in individual cryogenic vials containing >5x105 cells.

  • Excellent viability
  • Each vial of Corning™ HUVEC-2 cells contain 5x105 cells that have been cryopreserved at passage number 2 in medium containing 10% DMSO.
  • Lot-to-lot consistency
  • Tested for the ability to produce a robust migratory response in the presence of VEGF and FBS.
  • Prescreened for Angiogenesis studies
  • Qualified for use in Corning™ BioCoat Angiogenesis Systems: Endothelial Cell Migration. May also be used in Corning™ BioCoat Endothelial Cell Invasion and Tube Formation assays.

  • Positive immunohistochemical staining for von Willebrand factor (vWf) and CD31 antigen
  • Positive Dil-Ac-LDL uptake
  • Negative mycoplasma, HIV-1, hepatitis B and C, bacteria, yeast, and fungi

TRUSTED_SUSTAINABILITY

Specifications

For Use With Corning™ BioCoat™ Angiogenesis System
For Use With (Application) Used as model for studying endothelial differentiation and modulation of endothelial tube formation by antiangiogenic agents
Product Type Human Umbilical Vein Endothelial Cells
Quantity >5 x 105 Cells
Source Human Umbilical Vein
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.