Learn More
Corning™ Insectagro™ Sf9 Serum-free/Protein-free Medium, 1X, With L-glutamine
Shop All Corning ProductsDescription
- Satisfies demands of Sf9 and Sf21 cultures
- Cultured in non-humidified, non-CO2 incubators at 27°C
- Display both monolayer and suspension culture qualities
- With their fast doubling times of 18 to 22 hrs, Sf9 cells are easily scaled up to large cultures using bioreactors
- Capable of expressing full-length proteins
- Have been reported to produce certain proteins up to 20% of total protein
Order Info
Liquid and powdered media are packaged in PET with tamper-evident seals; shipped at ambient temperature. Certificates of Analysis for each lot are available from Mediatech upon request.
Compliance
Liquid products are tested for sterility according to FDA Guidelines for the Manufacture of In Vitro Diagnostic Products (January 1994) for sterile labeling.
Specifications
Specifications
| Concentration | 1 X |
| Form | Liquid |
| Product Type | Insect Cell Serum Free Medium (SFM), Protein-free |
| Sterility | Sterile |
| With Additives | Glutamine |
| For Use With (Application) | Insect cell culture |
| Quantity | 6 x 500 mL |
| Endotoxin Level | 0.1364 EU/ mL |
| pH | 6.7 |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.