Learn More
Corning™ GLS80 Linerless Screw Caps
Catalog No. 13700415 Shop All Corning Products
Change view
Click to view available options
Color:
Orange
Catalog No. Color
13-700-415 Orange
1 options
Catalog No. 13-700-415 Supplier Corning™ Supplier No. 139780LTC
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Polypropylene caps with linerless plug seal.
  • Orange polypropylene caps are autoclavable at 121°C
  • GLS80 thread
  • Linerless plug seal design for efficient sealing
TRUSTED_SUSTAINABILITY
Specifications
Closure Size GLS80
Product Type Screw Cap
Autoclavable Autoclavable
Material Polypropylene
Color Orange
No. per Pack 20/Pk.
No. per Case 20/Cs.
Sterility Non-sterile
WARNING: Cancer - www.P65Warnings.ca.gov